Product Name
Atto 488 DPPE suitable for fluorescence
Product Number
44039
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Red to Dark Red
APPEARANCE (FORM)Powder or Crystals
PURITY (HPCE AREA %)≥ 80.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 MG/ML IN CHCl3/MEOH 4:1 %(v/v)
EMISSION MAXIMUM527.0 ± 5.0 nm
REMARKS ON LUMINESCENCEEXC. 507 NM IN CHCl3/MEOH 4:1 %(v/v)
RECOMMENDED RETEST PERIOD36 MONTHS