Product Name
TRIS acetate-EDTA Buffer solution for Northern and Southern blotting, solution
Product Number
65497
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
DENSITY D20/40.995 - 1.004
REFRACTIVE INDEX N20/D1.332 - 1.336
PH-TEST8.1 - 8.5
PH (METHOD)NEAT, 25C
EXTRANEOUS ACTIVITIESDNASES, RNASES, PROTEASES, PHOSPHATASES NOT DETECTABLE