Product Name
12-(7-Nitrobenzofurazan-4-ylamino)dodecanoic acid suitable for fluorescence
Product Number
72963
Product Brand
sigma
CAS Number
96801-39-7
Molecular Weight
378.42
Test
Specification
APPEARANCE (COLOR)Yellow to Very Dark Yellow and Orange to Very Dark Orange and Brown to Very Dark Brown and Yellow-Orange and Yellow-Brown and Orange-Brown
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM530 ± 5 nm
REMARKS ON LUMINESCENCEEx. 466 nm in Methanol