Product Name
Atto 594 azide BioReagent, suitable for fluorescence, ≥80.0% (HPCE)
Product Number
72998
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Light Purple to Dark Purple
APPEARANCE (FORM)Powder or Solid
PURITY (HPCE AREA %)≥ 80.0 %
SOLUBILITY (COLOR)for information
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMF
EMISSION MAXIMUM627 ± 10 nm
REMARKS ON LUMINESCENCEEx. 601 nm in 0.1 M phosphate pH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS