Product Name
Atto 532 NHS ester BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Product Number
88793
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)PINK TO DEEP RED
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
DEGREE OF COUPLING≥ 80 %
EMISSION MAXIMUM551 ± 5 nm
REMARKS ON LUMINESCENCEEX. 530 NM IN 0.1 M PHOSPHATE PH 7.0
RECOMMENDED RETEST PERIOD36 MONTHS