Product Name
Blue Dextran average mol wt 5,000
Product Number
90008
Product Brand
sigma
CAS Number
87915-38-6
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue to Dark Blue
APPEARANCE (FORM)Powder
MOLECULAR NUMBERNUMBER AVERAGE MOLECULAR WEIGHT (MN): REPORT RESULT
MOLECULAR WEIGHT DETERMINATIONWEIGHT AVERAGE MOLECULAR WEIGHT (MW): 4000 - 6000
LOSS ON DRYING≤ 10 %
MISCELLANEOUS TESTSLOADING: 0.01 - 0.08 MMOL/G