Product Name
RNAstable® 96-well plate, for room temp preservation of RNA
Product Number
90220-001
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Functional testing-stability tested by extreme heat conditionsRNA stable sample remain stable at 60°C
Intrinsic Nuclease activityRNAstable sample do not exhibit intrinsic nuclease acitivity