Product Name
Pyrroloquinoline quinone ≥95.0% (HPLC)
Product Number
d7783
Product Brand
sigma
CAS Number
72909-34-3
Molecular Weight
330.21
Test
Specification
APPEARANCERED TO DEEP RED POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY YELLOWISH GREEN TO BROWN AT 10 MG/ML OF 1 M SODIUM HYDROXIDE
WATER CONTENT BY KARL FISCHERNOT MORE THAN 15%
IR SPECTRUMCONSISTENT WITH STRUCTURE
PURITY BY HPLCNOT LESS THAN 97.0%
DOCUMENT #D7783/09/19/07