Product Name
Anti-C20ORF27 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Number
hpa047483
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Protein Arraypass
Immunohistochemistry Dilution1:50 - 1:200
Immunohistochemistry Antibody Retrieval MethodHIER pH6
Concentration0.05 mg/mL