Product Name
Nortriptyline hydrochloride ≥98% (TLC), powder
Product Number
n7261
Product Brand
sigma
CAS Number
894-71-3
Molecular Weight
299.84
Test
Specification
Appearance (Color)White to Off-White
Appearance (Form)Powder
Solubility (Color)Colorless to Light Yellow
Solubility (Turbidity)Clear
 at 100 mg/ml in EtOH
Carbon74.2 - 78.0 %
Nitrogen4.4 - 5.0 %
Purity (TLC)≥98 %
Recommended Retest Period------------------------------
 5 years