Product Name
Tin(II) chloride dihydrate suitable for cell culture, ≥95%
Product Number
s9262
Product Brand
sigma
CAS Number
10025-69-1
Molecular Weight
225.65
Test
Specification
APPEARANCEWHITE TO WHITE WITH A YELLOW CAST POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY COLORLESS TO VERY FAINT YELLOW SOLUTION AT 100MG/ML IN 6M HYDROCHLORIC ACID
PURITY BY REDOX TITRATIONNLT 95%
CELL CULTURE TESTPASS