Product Name
Anti-C1QC antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
Product Number
sab1401035
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Western Blotting (Tissue lysate)Antibody concentration: 1 - 5 ug/ml. Specific band of ~25.77 kDa using human spleen lysate.
Western Blotting (Transfected lysate)Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~25.80 kDa using C1QC transfected 293T lysate.
Detection AntibodySuitable as a detection antibody in sandwich ELISA assay.