Product Name
Anti-SKIV2L2 antibody produced in rabbit affinity isolated antibody
Product Number
sab2102160
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
Concentration0.5-1 mg/ml
ELISANot Verified
Western BlottingAntibody dilution: 1 µg/ml
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified