Product Name
EG-VEGF human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
srp3029
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
ColorReport result
Purity by SDS-Page≥98%
Purity by HPLC≥98%
Endotoxin Level≤1 EU/µg