Product Name
IL-2 human recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Product Number
srp6170
Product Brand
sigma
CAS Number
110942-02-4
Molecular Weight
Test
Specification
AppearanceReport Result
Purity by SDS-PAGE≥ 95 %
Biological ActivityThe specific activity was determined by the dose-dependent stimulation of the proliferation of mouse CTLL-2 cells (mouse cytotoxic T cell line)