Product Name
Isobutyryl-L-carnitine-(N,N,N-trimethyl-d9) analytical standard
Product Number
08196
Product Brand
supelco
CAS Number
Molecular Weight
240.34
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
IDENTIFICATION (LC-MS)Identity Confirmed
REMARKS ON LC-MSDegree of Labelling ≥ 99 %
SPECIFIC ROTATION (20/D)-24.0 ± 2.0 Degrees
CONCENTRATIONc=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS