MilliporeSigma
Calcium carbonate precipitated (≤ 0.0001% Al), EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, FCC, E 170

Calcium carbonate

Calcium carbonate precipitated (≤ 0.0001% Al), EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, FCC, E 170
Synonyms:
Calcium carbonate
Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Special Grade
Technique

Products

Showing 1-20 of 31
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2