MilliporeSigma
n-Heptane for analysis EMPARTA®

Heptane

n-Heptane for analysis EMPARTA®
Synonym(s):
Heptane
Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20

Products

Shipping
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique