MilliporeSigma
Aluminum oxide for the determination of hydrocarbons

Aluminum oxide

Aluminum oxide for the determination of hydrocarbons
Synonym(s):
Alumina, Aluminium oxide 90 active neutral, Aluminium oxide 90 standardized, Aluminium oxide 90 active acidic (0.063-0.200 mm)
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Separation Technique
Quality Segment
Application
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-20 of 29
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2