MilliporeSigma
L-Serine ReagentPlus®, ≥99% (HPLC)

L-Serine

L-Serine ReagentPlus®, ≥99% (HPLC)
Synonym(s):
(S)-2-Amino-3-hydroxypropionic acid, L-Serine, (S)-(+)-Serine
Linear Formula:
HOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
105.09

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique