MilliporeSigma
L-Valine reagent grade, ≥98% (HPLC)

L-Valine

L-Valine reagent grade, ≥98% (HPLC)
Synonyms:
(S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid, L-Valine, (S)-(+)-Valine
Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique