MilliporeSigma
Starch from corn for use with Total Dietary Fiber Control Kit, TDF-C10

Starch from potato

Starch from corn for use with Total Dietary Fiber Control Kit, TDF-C10
Synonym(s):
Corn starch, Starch from maize, Starch (from wheat)
Linear Formula:
(C6H10O5)n
CAS Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Physical Form
Agency
Quality Segment
Application
Technique