MilliporeSigma
Glycogen from mussel, Mytilus genus for DNA precipitations

Glycogen from oyster

Glycogen from mussel, Mytilus genus for DNA precipitations
Synonym(s):
Glycogen, Molecular Biology Grade, Mussel
Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Physical Form
Purity
Research Area
Tissue
Quality Segment
Application
Package Size