1,2-Didecanoyl-sn-glycero-3-phosphocholine ≥99% (TLC)

1,2-Didecanoyl-sn-glycero-3-phosphocholine

1,2-Didecanoyl-sn-glycero-3-phosphocholine ≥99% (TLC)
Synonyms:
Didecanoyllecithin, 3-sn-Phosphatidylcholine, 1,2-didecanoyl, (7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxodecyl)oxy]-3,5,9-trioxa-4-phosphanonadecan-1-aminium 4-oxide, inner salt, PC(10:0/10:0), 1,2-Dicaprinoyl-sn-glycero-3-phosphocholine, L-α-Phosphatidylcholine, didecanoyl
Empirical Formula (Hill Notation):
C28H56NO8P
CAS Number:
Molecular Weight:
565.72

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
1,2-Didecanoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.