Skip to Content
MilliporeSigma
1,2-Phenylenediamine GR for analysis

1,2-Phenylenediamine

1,2-Phenylenediamine GR for analysis
Synonym(s):
1,2-Phenylenediamine
Linear Formula:
1,2-(NH2)2C6H4
CAS Number:
Molecular Weight:
108.14

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing