MilliporeSigma
L-Aspartic acid reagent grade, ≥98% (HPLC)

L-Aspartic acid

L-Aspartic acid reagent grade, ≥98% (HPLC)
Synonym(s):
(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid, L-Aspartic acid, (S)-(+)-Aspartic acid
Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique