MilliporeSigma
Sodium acetate trihydrate indifferent to potassium permanganate for analysis EMPARTA® ACS

Sodium acetate trihydrate

Sodium acetate trihydrate indifferent to potassium permanganate for analysis EMPARTA® ACS
Synonym(s):
Acetic acid sodium salt trihydrate, Sodium acetate trihydrate
Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08

Products

Shipping
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing