MilliporeSigma
Potassium phosphate monobasic NIST® SRM® 2186I

Potassium phosphate monobasic

Potassium phosphate monobasic NIST® SRM® 2186I
Synonyms:
Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate, Potassium phosphate monobasic
Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-20 of 28
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2