1,3-Diphenylacetone p-tosylhydrazone 98%

1,3-Diphenylacetone p-tosylhydrazone

1,3-Diphenylacetone p-tosylhydrazone 98%
Synonyms:
1,3-Diphenyl-2-propanone p-tosylhydrazone, 1,3-Diphenyl-2-propanone tosylhydrazone
Linear Formula:
CH3C6H4SO2NHN=C(CH2C6H5)2
CAS Number:
Molecular Weight:
378.49
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
1,3-Diphenylacetone p-tosylhydrazone, 98%

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.