MilliporeSigma
Iodomethane contains copper as stabilizer, ReagentPlus®, 99%

Iodomethane

Iodomethane contains copper as stabilizer, ReagentPlus®, 99%
Synonym(s):
Methyl iodide, Iodomethane
Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade