MilliporeSigma
N,N-Bis(2-hydroxyethyl) glycine (BICINE) buffer substance

BICINE

N,N-Bis(2-hydroxyethyl) glycine (BICINE) buffer substance
Synonyms:
N,N-Bis(2-hydroxyethyl)glycine, Bicine, ULTROL® Grade, N,N-Bis(2-hydroxyethyl) glycine
Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17

Products

Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application