MilliporeSigma
di-Sodium hydrogen phosphate dihydrate EMPLURA®

Sodium phosphate dibasic dihydrate

di-Sodium hydrogen phosphate dihydrate EMPLURA®
Synonym(s):
sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sodium phosphate dibasic dihydrate
Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99