MilliporeSigma
Aspirin meets USP testing specifications

Acetylsalicylic acid

Aspirin meets USP testing specifications
Synonyms:
2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin, Acetylsalicylic acid
Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
EC Number:

Products

Shipping
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
pK Value
Purity
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing