MilliporeSigma
Manganese(II) chloride tetrahydrate BioUltra, for molecular biology, ≥99.0% (KT)

Manganese(II) chloride tetrahydrate

Manganese(II) chloride tetrahydrate BioUltra, for molecular biology, ≥99.0% (KT)
Synonym(s):
Manganese(II) chloride tetrahydrate
Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique