Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
19-Norethindrone acetate
(1)
19-Norethindrone acetate
Synonyms:
19-Norethisterone 17-acetate, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one 17-acetate, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one acetate, 17α-Ethynyl-19-nortestosterone 17β-acetate
Empirical Formula (Hill Notation):
C
22
H
28
O
3
CAS Number:
51-98-9
Molecular Weight:
340.46
EC Number:
200-132-0
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
46527
19-Norethindrone acetate, VETRANAL
®
, analytical standard
Expand
View Pricing