MilliporeSigma
1-Methyl-2-Pyrrolidone for Peptide Synthesis

1-Methyl-2-Pyrrolidone

1-Methyl-2-Pyrrolidone for Peptide Synthesis
Synonyms:
1-Methyl-2-Pyrrolidone,, 1-Methyl-2-Pyrrolidone
Linear Formula:
(CH3)-NC(=O)CH2CH2CH2-
CAS Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
1-Methyl-2-Pyrrolidone, Biosynthesis OmniSolv®
1-Methyl-2-Pyrrolidone, OmniSolv®
1-Methyl-2-Pyrrolidone, Meets ACS Specifications GR ACS
1-Methyl-2-Pyrrolidone, for Peptide Synthesis