MilliporeSigma
1-Naphthylacetic acid 1 mg/mL, BioReagent, suitable for plant cell culture

1-Naphthylacetic acid

1-Naphthylacetic acid 1 mg/mL, BioReagent, suitable for plant cell culture
Synonyms:
α-Naphthaleneacetic acid Free acid, 1-Naphthaleneacetic acid, NAA, 1-Naphthylacetic acid
Linear Formula:
C10H7CH2CO2H
CAS Number:
Molecular Weight:
186.21

Products

Shipping
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Sterilization Method
Technique

Products

Showing 1-2 of 2