1-O-Octadecyl-2-O-methyl-sn-glycero-3-phosphorylcholine ≥99% (TLC), waxy solid

1-O-Octadecyl-2-O-methyl-sn-glycero-3-phosphorylcholine

1-O-Octadecyl-2-O-methyl-sn-glycero-3-phosphorylcholine ≥99% (TLC), waxy solid
Synonyms:
ET-18-OCH3
Empirical Formula (Hill Notation):
C27H58NO6P
Molecular Weight:
523.73

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
1-O-Octadecyl-2-O-methyl-sn-glycero-3-phosphorylcholine, ≥99% (TLC), waxy solid

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.