MilliporeSigma
1-Pyrenedecanoic acid suitable for fluorescence, ≥98.0% (HPLC)

1-Pyrenedecanoic acid

1-Pyrenedecanoic acid suitable for fluorescence, ≥98.0% (HPLC)
Synonyms:
10-(1-Pyrenyl)decanoic acid
Empirical Formula (Hill Notation):
C26H28O2
CAS Number:
Molecular Weight:
372.50

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
1-Pyrenedecanoic acid, suitable for fluorescence, ≥98.0% (HPLC)