(1S,2S,4R)-(+)-Limonene-1,2-diol ≥97.0% (GC)

(1S,2S,4R)-(+)-Limonene-1,2-diol

(1S,2S,4R)-(+)-Limonene-1,2-diol ≥97.0% (GC)
Synonyms:
(1S,2S,4R)-(+)-p-Menth-8-en-1,2-diol, (1S,2S,4R)-(+)-4-Isopropenyl-1-methylcyclohexan-1,2-diol, (1S,2S,4R)-8-p-Menth-8-ene-1,2-diol, (+)-1-Hydroxyneodihydrocarveol, (+)-(1S,2S,4R)-Limonene glycol
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
(1S,2S,4R)-(+)-Limonene-1,2-diol, ≥97.0% (GC)