MilliporeSigma
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for GC derivatization, LiChropur™, contains 1% TMCS, 99% (excluding TMCS)

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane for GC derivatization, LiChropur™, contains 1% TMCS, 99% (excluding TMCS)
Synonym(s):
BSTFA + TMCS, BSTFA, Bis(trimethylsilyl)trifluoroacetamide
Linear Formula:
CF3C[=NSi(CH3)3]OSi(CH3)3
CAS Number:
Molecular Weight:
257.40