MilliporeSigma
Cycloheximide PESTANAL®, analytical standard

Cycloheximide

Cycloheximide PESTANAL®, analytical standard
Synonyms:
3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide, Actidione, Naramycin A, Cycloheximide
Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique