2-Hydroxybenzaldehyde for synthesis

2-Hydroxybenzaldehyde

2-Hydroxybenzaldehyde for synthesis
Synonyms:
2-Hydroxybenzaldehyde
Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
2-Hydroxybenzaldehyde, for synthesis

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.