MilliporeSigma
2-Methoxyphenylacetonitrile 98%

2-Methoxyphenylacetonitrile

2-Methoxyphenylacetonitrile 98%
Synonyms:
2-Methoxybenzyl cyanide
Linear Formula:
CH3OC6H4CH2CN
CAS Number:
Molecular Weight:
147.17
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
2-Methoxyphenylacetonitrile, 98%