2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium salt ≥75% (TLC)

2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium salt

2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium salt ≥75% (TLC)
Synonyms:
1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phospho-L-serine sodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine sodium salt, POPS-Na, (2S,8R,18Z)- 2-Amino-5-hydroxy-10-oxo-8-[[(1-oxohexadecyl)oxy]methyl]-4,6,9-trioxa-5-phosphaheptacos-18-enoic acid 5-oxide monosodium salt, PS(16:0/18:1), 3-sn-Phosphatidyl-L-serine, 2-oleoyl-1-palmitoyl sodium salt, PS(16:0/18:1(9Z))
Empirical Formula (Hill Notation):
C40H75NNaO10P
CAS Number:
Molecular Weight:
783.99

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium salt, ≥75% (TLC)

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.