3,3-BIS-CHLOROMETHYL-CYCLOPROPANE-1,1,2,2-TETRACARBONITRILE AldrichCPR

3,3-BIS-CHLOROMETHYL-CYCLOPROPANE-1,1,2,2-TETRACARBONITRILE

3,3-BIS-CHLOROMETHYL-CYCLOPROPANE-1,1,2,2-TETRACARBONITRILE AldrichCPR
Linear Formula:
C9H4Cl2N4
Molecular Weight:
239.065

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
3,3-BIS-CHLOROMETHYL-CYCLOPROPANE-1,1,2,2-TETRACARBONITRILE, AldrichCPR
3,3-BIS-CHLOROMETHYL-CYCLOPROPANE-1,1,2,2-TETRACARBONITRILE, AldrichCPR

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.