MilliporeSigma
DAPI for nucleic acid staining

DAPI

DAPI for nucleic acid staining
Synonym(s):
4′,6-Diamidino-2-phenylindole dihydrochloride, 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride, DAPI Stain, DAPI, Dihydrochloride
Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25

Products

Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique