3,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3 98 atom % 15N, 99 atom % 13C, 98% (CP)

3,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3

3,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3 98 atom % 15N, 99 atom % 13C, 98% (CP)
Synonyms:
3,7-Dimethyl-2,6,8-trihydroxypurine-13C4,15N3 (2,4,5,6-13C4, 1,3,9-15N3)
Empirical Formula (Hill Notation):
13C4C3H815N3NO3
Molecular Weight:
203.11

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
Product Number
Product Description
Pricing
3,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3, 98 atom % 15N, 99 atom % 13C, 98% (CP)

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.