Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
3′-Demethyl-3′-N-[(4-Methylphenyl)Sulfonyl]Azithromycin Pharmaceutical Analytical Impurity (PAI)

3′-Demethyl-3′-N-[(4-Methylphenyl)Sulfonyl]Azithromycin

3′-Demethyl-3′-N-[(4-Methylphenyl)Sulfonyl]Azithromycin Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
(N-((2S,4S,6R)-2-(((2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4- methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-15-oxo-1-oxa-6-azacyclopentadecan11-yl)oxy)-3-hydroxy-6-methyltetrahydro-2H-pyran-4-yl)-N,4-dimethylbenzenesulfonamide)
Empirical Formula (Hill Notation):
C44H76N2O14S
CAS Number:
Molecular Weight:
889.14

Products

Product Category
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1