MilliporeSigma
(±)-3-Methyl-2-oxovaleric acid sodium salt

(±)-3-Methyl-2-oxovaleric acid sodium salt

(±)-3-Methyl-2-oxovaleric acid sodium salt
Synonyms:
(±)-Sodium 3-methyl-2-oxovalerate, Ketoisoleucine sodium salt, DL-α-Keto-β-methylvaleric acid sodium salt, 3-Methyl-2-oxopentanoic acid sodium salt
Linear Formula:
CH3CH2CH(CH3)COCOONa
CAS Number:
Molecular Weight:
152.12
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(±)-3-Methyl-2-oxovaleric acid sodium salt